missing translation for 'onlineSavingsMsg'
Learn More

Jaytec Glass™ Borosilicate Glass Butyrometer Tube

The Borosilicate Glass Butyrometer Tube is used to determine the fat content of dairy samples using the Gerber method.

43.15 € - 45.35 €

Specifications

Material Borosilicate Glass
For Use With (Application) Determination of fat content using Gerber method
View More Specs

Products 2
Product Code Brand Type Capacity (Metric) Range Graduations Price Quantity & Availability  
Product Code Brand Type Capacity (Metric) Range Graduations Price Quantity & Availability  
12388955
View Documents
Jaytec Glass™
MP890
Cheese Butyrometer 3 g 0 to 40% 1%
45.35 €
Each
Estimated Shipment: 03-03-2025
to see stock.
Add to basket
 
12378955
View Documents
Jaytec Glass™
MP893
Milk Butyrometer 10.94 mL 0 to 8% 0.1%
43.15 €
Each
In Stock
to see stock.
Edge
Add to basket
 
Description

Description

  • Borosilicate glass
  • Capacity and tolerances in accordance with BS ISO 488:2008 (formerly BS 696)
  • Graduated round stem
Specifications

Specifications

Borosilicate Glass
Determination of fat content using Gerber method
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.