Learn More
BRAND™ BLAUBRAND™ Borosilicate Glass Centrifuge Tube
Withstands RCF up to 700. Design and accuracy meet ASTM (American Society for Testing and Materials) specifications.
Brand:  BRAND™ 362338
88.92 EUR valid until 2025-01-31
Use promo code "23894" to get your promotional price.
 View more versions of this product
Product Code. 11894050
  • 101.00 € / Pack of 2
In Stock
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Withstands RCF up to 700.
  • Borosilicate glass construction (Boro 3.3)
  • Cylindrical with conical bottom
TRUSTED_SUSTAINABILITY
Specifications
Specifications
100 mL
167 mm
96 mm
700 x g
2
Conical
Centrifuge Tube