Learn More
Cytiva Borosilicate Glass Chromatographic Tubes
Resistant to aqueous solutions and solvents used in liquid chromatography
Brand:  Cytiva 19-5036-01
 View more versions of this product
Product Code. 10231110
  • 151.00 € / Each
Estimated Shipment: 23-04-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
Not resistant to acetone, ketones, chlorinated hydrocarbons, aliphatic esters, phenol, NaOH, > 10% HCl, > 5% acetic acid, or strong mineral acid.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
For C 10/20 Columns
10 mm
Borosilicate Glass
60°C
Chromatography Tubes
200 mm
14.5 psi