Learn More
Kimble Chase™ Borosilicate Glass Chromatography Column with Fritted Disc and PTFE Stopcock Plug
Gravity chromatography column manufactured from medium wall tubing with 821000 PTFE stopcock
610.00 €
Specifications
Type | Chromatography Column |
---|---|
For Use With (Application) | Ion exchange chromatography |
Quantity | 1 Each |
USP Type | Type I |
Material | Borosilicate Glass |
Description
- 40-60 micron porosity glass frit at the lower end to act as a support for column packing material
Specifications
Chromatography Column | |
1 Each | |
Borosilicate Glass |
Ion exchange chromatography | |
Type I | |
With fritted disc and PTFE stopcock plug |