missing translation for 'onlineSavingsMsg'
Learn More

Cytiva Borosilicate Glass Columns

Easy to use, general purpose glass columns

Brand:  Cytiva 18-1115-07

 View more versions of this product

Product Code. 11340242

  • 5292.00 € / Each
Estimated Shipment: 14-08-2026
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

INdEX columns are easy-to-use, general purpose, glass columns well-suited for applications such as process development and diagnostics production. These columns are characterized by their simple design, simplicity of operation and the novel, axial compression packing method that yields a densely packed bed in under 10 minutes.
  • Scalable from inner diameters of 70 to 200mm and bed volumes from 0.1L to 25L.
  • Pressure rating: 3 bar.
  • For use with an array of techniques and chromatography media, especially Sepharose Fast Flow.
  • Proven distribution system.
  • Dynamic axial compression yields densely packed, highly efficient beds.
  • Materials include electropolished stainless steel, borosilicate glass, and polymers.
  • Packing devices for longer bed heights.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Process development and small-scale production
70 mm
3 bar
500 mm
Polyamid and Polypropylene
1 Pack
Column
10 and 23 μm
Borosilicate Glass
25mm TC
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.