Learn More
BRAND™ BLAUBRAND™ Compact Automatic Burette with Plastic Bottle
Fast to dismantle and easy to clean. All individual components are replaceable.
Brand:  BRAND™ 23821
132.90 EUR valid until 2025-01-31
Use promo code "23894" to get your promotional price.
Product Code. 10778785
  • 153.00 € / Each
Estimated Shipment: 09-01-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Borosilicate glass (Boro 3.3) construction
  • Class AS, DE-M marking
  • Graduations in blue
  • Includes 1000 mL LDPE bottle with pedestal, PVC filling tube and detachable PTFE stopcock
  • DIN EN ISO 385
  • Calibrated to deliver (TD, Ex)
  • Supplied with a batch number and an accompanying batch certificate in the original packaging
  • On request, they are also available with an individual certificate, USP individual certificate or DAkkS calibration certificate
  • Support/clamp not included
  • TRUSTED_SUSTAINABILITY
    Specifications
    Specifications
    Clear
    Borosilicate Glass
    PE Bottle
    Schellbach Buret
    DIN EN ISO 385, USP, DIN EN ISO 385
    50 mL
    0.1 mL
    PTFE
    Class AS