Learn More
Borosilicate Glass Conical Form Beakers with Wide Mouth
Product Code. 12707836
Click to view available options
Capacity (Metric):
150 mL
250 mL
500 mL
Use Borosilicate Glass Conical Form Beakers with Wide Mouth in the laboratory for measuring liquids and chemicals during experimentation. The conical beakers are available in three metric capacities.
  • Borosilicate Glass
  • Conical
  • Graduated
  • Wide Mouth
Specifications
Capacity (Metric) 500 mL
Material Borosilicate Glass
Includes Spout
Quantity 1 Ea.
Type Beaker