Learn More
Lenz Laborglasinstrumente™ Separating Funnels, with PTFE Plug and Scale
For use in liquid-liquid extractions to separate components. Lenz Laborglasinstrumente™ Separating Funnels, with PTFE Plug and Scale are manufactured from durable DURAN™ borosilicate glass.
Brand: Lenz Laborglasinstrumente™ 04006137
This item is not returnable.
View return policy
Description
- With scale (approx. volume/mL)
- With PTFE plug
- With NS socket and PE stopper
- With screwthread retaining device
- Corrosion-free and resistant to thermal shock
Specifications
100 mL | |
Borosilicate Glass | |
2Pack | |
Polyethylene |
Product Content Correction
Product Title
Lenz Laborglasinstrumente™ Separating Funnels, with PTFE Plug and Scale > Capacity: 100mL
Spot an opportunity for improvement?Share a Content Correction