Learn More
Lenz Laborglasinstrumente™ Separating Funnels, with PTFE Plug and Scale
For use in liquid-liquid extractions to separate components. Lenz Laborglasinstrumente™ Separating Funnels, with PTFE Plug and Scale are manufactured from durable DURAN™ borosilicate glass.
Brand:  Lenz Laborglasinstrumente™ 04006173
 View more versions of this product
Product Code. 11836084
  • 190.00 € / Each
Estimated Shipment: 23-01-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • With scale (approx. volume/mL)
  • With PTFE plug
  • With NS socket and PE stopper
  • With screwthread retaining device
  • Corrosion-free and resistant to thermal shock
Specifications
Specifications
2000 mL
Borosilicate Glass
Polyethylene