Learn More
Lenz Laborglasinstrumente™ Separating Funnels, with PTFE Plug and Scale
For use in liquid-liquid extractions to separate components. Lenz Laborglasinstrumente™ Separating Funnels, with PTFE Plug and Scale are manufactured from durable DURAN™ borosilicate glass.
Brand:  Lenz Laborglasinstrumente™ 04006170
 View more versions of this product
Product Code. 11852672
  • 170.00 € / Pack of 2
Estimated Shipment: 17-12-2024
to see stock.
Explore more special offers
Add to basket
Description
Description
  • With scale (approx. volume/mL)
  • With PTFE plug
  • With NS socket and PE stopper
  • With screwthread retaining device
  • Corrosion-free and resistant to thermal shock
Specifications
Specifications
1000 mL
Borosilicate Glass
2Pack
Polyethylene