missing translation for 'onlineSavingsMsg'
Learn More

Eberbach Glass Container with Lid

Glass Container includes a cover and blades.

Brand:  Eberbach 04243-28

Product Code. 11974908

  • 877.00 € / Each
Estimated Shipment: 12-02-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • 1 year warranty.
Specifications

Specifications

Container
3.75 in.
165.1 mm
Metal Screw Cover
30 to 250 mL
95.2 mm
Borosilicate Glass
1 Ea.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers