missing translation for 'onlineSavingsMsg'
Learn More

BRAND™ Borosilicate Glass Cover Glasses

Clear borosilicate glass construction with excellent chemical resistance.

Brand:  BRAND™ 470055

96.80 EUR valid until 2025-12-31
Use promo code "25339" to get your promotional price.


Product Code. 15212469

  • 129.00 € / Pack of 2000
In Stock
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • Clear borosilicate glass
  • Hydrolytic class 1
  • Thickness No. 1 (0.13 to 0.17 mm)
  • Refractive index ne = 1.52 ± 0.01
  • Abbe number ve = 56.5 ± 0.5.
  • Full automatic production guarantees ready-for-use quality: clean, dust and grease-free, distortion-free
  • Flatness within ± 3 μm.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Coverslip
Borosilicate Glass
22 x 22 mm
2000
Square
#1 (0.13 to 0.16 mm)
Microscope slides
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.