Learn More
Karl Hecht™ Assistent™ Reduction and Expansion Adapters
For the quick and secure connection of glass joints in the laboratory. Karl Hecht™ Assistent™ Reduction and Expansion Adapters are manufactured from durable DURAN™ borosilicate 3.3 glass.
Brand: Karl Hecht™ 42387008
This item is not returnable.
View return policy
Description
- Solid rim
- Acc. to DIN 12257
- Reinforced ground joint
Specifications
Borosilicate Glass | |
Duran sleeve | |
1 Each | |
Socket size: 19/26, Cone size: 14/23 |