missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Quickfit Jointed Ware™ Borosilicate Glass Florentine Flask
SciLabWare™ Quickfit Jointed Ware™ Borosilicate Glass Florentine Flask is ideal for use with rotary evaporators.
Brand: Quickfit FD2L/4RE
93.15 EUR valid until 2024-12-31
Use promo code "23894" to get your promotional price.
This item is not returnable.
View return policy
Description
SciLabWare™ Quickfit Jointed Ware™ Borosilicate Glass Florentine Flask is ideal for use with rotary evaporators.
- Ideal for use with rotary evaporators
- Large surface area for rapid evaporation
- Robust heavy wall construction for added strength
- Shape allows easy removal of solid residues
Specifications
2000mL | |
158mm | |
245mm | |
Florentine Flask |
2000mL | |
Rotary evaporator | |
Borosilicate Glass |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction