Learn More
Lenz Laborglasinstrumente™ Gas Washing Bottles without Head
Provides an inexpensive but effective method for the washing and drying of gases. Lenz Laborglasinstrumente™ Gas Washing Bottles without Head are manufactured from durable DURAN™ borosilicate glass.
Brand:  Lenz Laborglasinstrumente™ 05515037
 View more versions of this product
Product Code. 10548681
  • 23.00 € / Each
Estimated Shipment: 12-02-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Acc. to DIN 12596, without head
  • With socket NS 29/32
  • Available in a range of capacities
  • Corrosion-free and resistant to thermal shock
Specifications
Specifications
100 mL
Gas Washing Bottle without Head
NS 29/32
DIN 12463
Borosilicate Glass
Socket
200 mm
1 Pack