Learn More
Fisherbrand™ Glass Circle Coverslips
Glass Circle Coverslips are available in two thicknesses and a variety of diameters. Packed in plastic boxes to prevent breakage.
4.14 € - 32.85 €
Product Code | Brand | Thickness (Metric) | Diameter (Metric) | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Thickness (Metric) | Diameter (Metric) | Price | Quantity & Availability | |||||
12313138
|
Fisherbrand™
MCR20016 |
#1 (0.13 to 0.16 mm) | 16 mm |
30.35 €
Pack of 200 |
In Stock
Log in to see stock. ![]() |
|||||
12323148
|
Fisherbrand™
MCR00016 |
#1.5 (0.16 to 0.19 mm) | 16 mm |
4.14 €
Pack of 100 |
In Stock
Log in to see stock. ![]() |
|||||
12333138
|
Fisherbrand™
MCR20022 |
#1 (0.13 to 0.16 mm) | 22 mm |
32.85 €
Pack of 200 |
In Stock
Log in to see stock. ![]() |
|||||
12323138
|
Fisherbrand™
MCR20019 |
#1 (0.13 to 0.16 mm) | 19 mm |
30.40 €
Pack of 200 |
In Stock
Log in to see stock. ![]() |
|||||
11507323
|
Fisherbrand™
MCR00022 |
#1.5 (0.16 to 0.19 mm) | 22 mm | N/A | ||||||
Description
These Glass Circle Coverslips are available in two thicknesses and a variety of diameters. Packed in plastic boxes to prevent breakage.
- Manufactured from clear soda glass
- Consistent in cut, thickness and dimensional accuracy
- Free from blisters and flaws
Specifications
Coverslip | |
Circle | |
Soda Lime Glass |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Product Title
Fisherbrand™ Glass Circle Coverslips