missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Corning™ Borosilicate Glass Pasteur Pipets
Manufactured with Borosilicate Glass for optimum visibility. Corning™ Borosilicate Glass Pasteur Pipets are ideal for rapid non-volumetric transfer work.
73.00 € - 81.26 €
Specifications
Diameter (Metric) Outer | 6.5 mm |
---|---|
Material | Borosilicate Glass |
For Use With (Application) | Ideal for rapid non-volumetric transfer work in bacteriology, immunology, hematology and serology studies, as well as blood bank and clinical chemistry procedures |
Quantity | 1000 Pack |
Tip Style | Round |
Product Code | Brand | Length (Metric) | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Length (Metric) | Price | Quantity & Availability | |||||
10323523
|
Corning™
7095D-9 |
229 mm |
81.26 €
Pack of 1000 |
Estimated Shipment: 28-02-2025
Log in to see stock. |
|||||
10494533
|
Corning™
7095D-5X |
146 mm |
73.00 €
Pack of 1000 |
Estimated Shipment: 28-02-2025
Log in to see stock. |
|||||
Description
- Available in convenient bulk-packs
- One-time use eliminates the danger of cross-contamination of valuable specimens or laboratory reagents
- Feature a constriction one inch below the top to facilitate plugging
- Ideal for work in bacteriology, immunology, hematology and serology studies
- Use for blood bank and clinical chemistry procedures
Specifications
6.5 mm | |
Ideal for rapid non-volumetric transfer work in bacteriology, immunology, hematology and serology studies, as well as blood bank and clinical chemistry procedures | |
Round | |
Single-use |
Borosilicate Glass | |
1000 Pack | |
Pasteur Pipet |