missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Wheaton™ Borosilicate Glass Pestle
Brand: Wheaton™ 358105
137.78 EUR valid until 2024-12-31
Use promo code "23894" to get your promotional price.
This item is not returnable.
View return policy
Description
Specifications
Autoclavable | |
Borosilicate Glass | |
For homogenizing connective tissue including heart, muscle and lung |
3 mL | |
Pestle | |
For 358107 (3mL) Tapered Tissue Grinder Set |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction