Learn More
Lenz Laborglasinstrumente™ Pointed Bottom Hollow Stoppers, Hexagonal Plate
For the safe and secure closure of flasks, bottles, tubing and other laboratory apparatus. Lenz Laborglasinstrumente™ Pointed Bottom Hollow Stoppers, Hexagonal Plate are manufactured from durable DURAN™ borosilicate glass.
16.00 € - 40.00 €
Specifications
Description Hollow
Material Borosilicate Glass
Product Type Hollow Stopper
Quantity 10/Pk.
View More Specs
Products 6
Product Code Brand Standard Taper Joint Price Quantity & Availability  
Product Code Brand Standard Taper Joint Price Quantity & Availability  
10495251
View Documents
Lenz Laborglasinstrumente™
01400010
Thread Size: 10/19 Cone NS
18.00 €
Pack of 10
In Stock
to see stock.
Edge
Add to basket
10769121
View Documents
Lenz Laborglasinstrumente™
01400012
Thread Size: 12/21 Cone NS
16.00 €
Pack of 10
Estimated Shipment: 12-12-2024
to see stock.
Add to basket
15320885
View Documents
Lenz Laborglasinstrumente™
01400019
Thread Size: 19/26 Cone NS
24.00 €
Pack of 10
In Stock
to see stock.
Add to basket
10550052
View Documents
Lenz Laborglasinstrumente™
01400024
Thread Size: 24/29 Cone NS
27.00 €
Pack of 10
In Stock
to see stock.
Add to basket
10493341
View Documents
Lenz Laborglasinstrumente™
01400029
Thread Size: 29/32 Cone NS
40.00 €
Pack of 10
In Stock
to see stock.
Add to basket
10261731
View Documents
Lenz Laborglasinstrumente™
01400014
Thread Size: 14/23 Cone NS
20.00 €
Pack of 10
In Stock
to see stock.
Add to basket
Description
Description
  • Acc. to DIN 12252, form C
  • Available in a range of cone sizes
  • Corrosion-free and resistant to thermal shock
Specifications
Specifications
Hollow
Borosilicate Glass
Hollow Stopper
10/Pk.