Learn More
BRAND™ Borosilicate Glass Powder Funnel
Manufactured from borosilicate glass. BRAND™ Borosilicate Glass Powder Funnel includes ground and fire-polished upper rim. Short, wide stem is optimal for funneling powder and other ground materials.
Brand:  BRAND™ 146517
 View more versions of this product
Product Code. 10181531
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Explore more special offers
Description
Description
  • Manufactured from borosilicate glass.
  • Ground and fire-polished upper rim.
  • Short, wide stem optimal for funneling powder and other ground materials.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
90 mm
25 mm
Borosilicate Glass