Learn More
Scientific Glass Labs™ Borosilicate Glass Powder Funnel
Manufactured from borosilicate glass. Scientific Glass Labs™ Borosilicate Glass Powder Funnel is less likely to clog or cake when funneling powdery substances.
5.05 € - 8.30 €
Product Code | Brand | Diameter (Metric) Stem | Diameter (Metric)Top | Length (Metric) Stem | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Diameter (Metric) Stem | Diameter (Metric)Top | Length (Metric) Stem | Price | Quantity & Availability | |||
12993591
|
Scientific Glass Labs™
FPH-240-070U |
16mm | 70mm | 25mm |
5.05 €
Each |
In Stock
Log in to see stock. |
|||
12903601
|
Scientific Glass Labs™
FPH-240-090X |
22mm | 100mm | 25mm |
8.30 €
Each |
Estimated Shipment: 22-01-2025
Log in to see stock. |
|||
12983591
|
Scientific Glass Labs™
FPH-240-050D |
13mm | 50mm | 20mm |
5.80 €
Each |
In Stock
Log in to see stock. |
|||
Description
- Manufactured from borosilicate glass.
- Short stem funnel
- Less likely to clog or cake when funneling powdery substances
Specifications
Borosilicate Glass |