Learn More
Lenz Laborglasinstrumente™ Borosilicate Glass Separating Funnels with PTFE Plug
Made of DURAN™ borosilicate glass. Lenz Laborglasinstrumente™ Borosilicate Glass Separating Funnels with PTFE Plug are used in liquid-liquid extractions to separate the components of a mixture into immiscible solvent phases of different densities.
Brand:  Lenz Laborglasinstrumente™ 04030170
 View more versions of this product
Product Code. 10102051
  • 410.00 € / Pack of 2
Estimated Shipment: 27-01-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • With NS socket and PE stopper
  • Without scale, with PTFE plug
  • With screwthread retaining device
  • With inscription label
Specifications
Specifications
1000 mL
29/32 NS
Polyethylene
Borosilicate Glass
6mm