missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Lenz Laborglasinstrumente™ Borosilicate Glass Separating Funnels with Solid Glass Plug
Lenz Laborglasinstrumente™ Borosilicate Glass Separating Funnels with Solid Glass Plug are used in liquid-liquid extractions to separate the components of a mixture into immiscible solvent phases of different densities.
191.00 € - 336.00 €
Product Code | Brand | Capacity (Metric) | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Capacity (Metric) | Price | Quantity & Availability | |||||
10090531
|
Lenz Laborglasinstrumente™
04030037 |
100 mL |
191.00 €
Pack of 2 |
Estimated Shipment: 02-01-2025
Log in to see stock. |
|||||
10213961
|
Lenz Laborglasinstrumente™
04030049 |
250 mL |
212.00 €
Pack of 2 |
Estimated Shipment: 02-01-2025
Log in to see stock. |
|||||
10729221
|
Lenz Laborglasinstrumente™
04030070 |
1000 mL |
336.00 €
Pack of 2 |
Estimated Shipment: 09-01-2025
Log in to see stock. |
|||||
10040381
|
Lenz Laborglasinstrumente™
04030058 |
500 mL |
242.00 €
Pack of 2 |
Estimated Shipment: 24-12-2024
Log in to see stock. |
|||||
Description
- Cylindrical shape with straight discharge tube
- With NS socket and PE stopper
- With solid glass plug
- With Screwthreade retaining device
- With scale
Specifications
Borosilicate Glass | |
Polyethylene |