missing translation for 'onlineSavingsMsg'
Learn More

Kimble Chase™ Borosilicate Glass Shell Vials

Use Kimble™ Borosilicate Glass Shell Vials for storage of dry contents.

Specifications

Color Clear
Material Borosilicate Glass
For Use With (Application) Ideal for Storing Dry Products
Type Shell Vial
Graduated Yes
View More Specs

Products 2
Product Code Brand Capacity (Metric) Price Quantity & Availability  
Product Code Brand Capacity (Metric) Price Quantity & Availability  
10052582
View Documents Promotion Details
Kimble Chase™
60931-12
2 mL N/A
N/A
11873133
View Documents Promotion Details
Kimble Chase™
60931-12
2 mL N/A
N/A
Description

Description

Use Kimble™ Borosilicate Glass Shell Vials for storage of dry contents.

  • Straight-sided
  • Clear N-51A borosilicate glass
  • Without caps
Specifications

Specifications

Clear
Ideal for Storing Dry Products
Yes
Borosilicate Glass
Shell Vial
Videos
SDS
Documents

Documents

Product Certifications