Learn More
Kimble Chase™ Borosilicate Glass Shell Vials, No Cap
Short-style shell vials are ideal for storing dry products
Specifications
No. per Pack 144
View More Specs
Description
Description
  • Plain top design
  • Lab pack quantities
  • Manufactured from 51 expansion borosilicate glass conforming to USP Type I and ASTM E438, Type I, Class B requirements
Specifications
Specifications
144