Learn More
Fisherbrand™ Soda Lime Glass Short Stem Funnel
Manufactured from soda lime glass. Fisherbrand™ Soda Lime Glass Short Stem Funnel is less likely to clog when funneling thicker substances.
Brand:  Fisherbrand™ 00793106
 View more versions of this product
Product Code. 10124352
  • 37.75 € / Pack of 10
In Stock
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Manufactured from Soda Lime glass.
  • Short stem funnel
  • Less likely to clog when funneling thicker substances.
Specifications
Specifications
55mm
Soda Lime Glass
Ideal for filtering applications in the laboratory
8mm
95 mm
60mm
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
One moment while we fetch your results.
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Fisherbrand™ Soda Lime Glass Short Stem Funnel > Top: 55mm dia.; Stem Length: 55mm