Learn More
Fisherbrand™ Borosilicate Glass Squat Form Heavy Duty Beakers Product Code.: 15496123
Add to basket
Fisherbrand™ Borosilicate Glass Squat Form Heavy Duty Beakers
Product Code. 15496123 Shop All Fisherbrand Products
600 mL
Click to view available options
Capacity (Metric):
150 mL
250 mL
400 mL
600 mL
1000 mL
2000 mL
You may only order up to the available stock. Where an alternative item has been identified it will be shown below
Product Code. 15496123
Add to basket
Add to basket
You may only order up to the available stock. Where an alternative item has been identified it will be shown below
Feature heavy wall construction for increased mechanical strength. Fisherbrand™ Borosilicate Glass Squat Form Heavy Duty Beakers are made of full 3.3 expansion borosilicate glass and feature an 'easy pour' spout.
  • Heavy wall construction for increased mechanical strength
  • Full 3.3 expansion borosilicate glass
  • Reinforced rim with 'easy pour' pouring spout
  • Graduated in white ceramic marking area
  • Uniform wall thickness for optimm optical clarity
TRUSTED_SUSTAINABILITY
Specifications
Capacity (Metric) 600 mL
Material Borosilicate Glass
Includes Spout
Quantity 10
Type Beaker
Graduated Yes
Scale White Ceramic Scale
Style Squat Form
Product Content Correction
Product Title
Fisherbrand™ Borosilicate Glass Squat Form Heavy Duty Beakers >