Learn More
Lenz Laborglasinstrumente™ Separating Funnels Acc. to Squibb, with PTFE Plug
For use in liquid-liquid extractions to separate components. Lenz Laborglasinstrumente™ Separating Funnels Acc. to Squibb, with PTFE Plug are manufactured from durable DURAN™ borosilicate glass.
Brand:  Lenz Laborglasinstrumente™ 04014137
 View more versions of this product
Product Code. 10567541
  • 90.00 € / Pack of 2
Estimated Shipment: 22-01-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Without scale
  • With NS socket and PE stopper
  • With screwthread retaining device
  • Corrosion-free and resistant to thermal shock
Specifications
Specifications
100 mL
Borosilicate Glass
2Pack
Polyethylene