Learn More
Lenz Laborglasinstrumente™ Borosilicate Glass Squibb Separating Funnels with PTFE Plug, Scale
Lenz Laborglasinstrumente™ Borosilicate Glass Squibb Separating Funnels with PTFE Plug, Scale are used in liquid-liquid extractions to separate the components of a mixture into immiscible solvent phases of different densities.
86.00 € - 148.00 €
Product Code | Brand | Capacity (Metric) | Price | Quantity & Availability | ||||||
---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Capacity (Metric) | Price | Quantity & Availability | ||||||
11822662
|
Lenz Laborglasinstrumente™
04016170 |
1000 mL |
148.00 €
Pack of 2 |
Estimated Shipment: 19-05-2025
Log in to see stock. |
||||||
10110551
|
Lenz Laborglasinstrumente™
04016149 |
250 mL |
103.00 €
Pack of 2 |
Estimated Shipment: 02-05-2025
Log in to see stock. |
||||||
11892652
|
Lenz Laborglasinstrumente™
04016137 |
100 mL |
86.00 €
Pack of 2 |
Estimated Shipment: 25-04-2025
Log in to see stock. |
||||||
11846164
|
Lenz Laborglasinstrumente™
04016173 |
2000 mL |
142.48 €
Each |
Estimated Shipment: 17-04-2025
Log in to see stock. |
||||||
11812662
|
Lenz Laborglasinstrumente™
04016158 |
500 mL |
132.08 €
Pack of 2 |
Estimated Shipment: 06-05-2025
Log in to see stock. |
||||||
11822652
|
Lenz Laborglasinstrumente™
04016128 |
50 mL |
90.00 €
Pack of 2 |
Estimated Shipment: 12-05-2025
Log in to see stock. |
||||||
Description
- With NS socket and PE stopper
- with PTFE plug
- with screwthread retaining device
- with scale (approx. volume /ml)
- material: borosilicate glass 3.3
Specifications
Borosilicate Glass | |
Polyethylene |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Product Title
Lenz Laborglasinstrumente™ Borosilicate Glass Squibb Separating Funnels with PTFE Plug, Scale