Learn More
Hamilton™ 7000 Series Knurled Hub Syringes
Modified Microliter syringes with a plunger-in-needle design.
Brand:  Hamilton™ DS88500
 View more versions of this product
Product Code. 10005590
  • 2010.00 € / Each
Explore more special offers
Description
Description
  • The only zero dead volume syringe available
  • Fine gauge tungsten plunger wire extends to the tip of the needle
  • Ensures that all solvent is expelled
Specifications
Specifications
Borosilicate Glass, Stainless Steel
Syringe
3
23 G
1/Ea.