Learn More
Cytiva Borosilicate Glass XK Chromatography Tubes
Can be used in aqueous solutions and nearly all organic solvents commonly used in liquid chromatography of macromolecules.
Brand:  Cytiva 19-0146-01
 View more versions of this product
Product Code. 10079619
  • 388.00 € / Each
In Stock
to see stock.
Explore more special offers
Add to basket
Description
Description
  • For use with many organic solvents except chlorinated hydrocarbons, acetone and other ketones, aliphatic esters and phenol
  • Solutions containing more than 10% NaOH, 10% HCl, other strong mineral acids or 5% acetic acid should not be used
TRUSTED_SUSTAINABILITY
Specifications
Specifications
26/70 Columns
26 mm
Borosilicate Glass
60°C
XK Column Tubes
70 cm
4°C