Learn More
Cytiva Borosilicate Glass XK Chromatography Tubes
Can be used in aqueous solutions and nearly all organic solvents commonly used in liquid chromatography of macromolecules.
Brand: Cytiva 19-0113-01
158.65 EUR valid until 2024-10-31
Use promo code "21692" to get your promotional price.
This item is not returnable.
View return policy
Description
- For use with many organic solvents except chlorinated hydrocarbons, acetone and other ketones, aliphatic esters and phenol
- Solutions containing more than 10% NaOH, 10% HCl, other strong mineral acids or 5% acetic acid should not be used
Specifications
16/40 Columns | |
16 mm | |
Borosilicate Glass | |
60°C |
XK Column Tubes | |
40 cm | |
4°C |