missing translation for 'onlineSavingsMsg'
Learn More

VITLAB™ Borosilicate Dr. Schilling Burette

Turn the burette stopcock easily to enable fine titration with the Vitlab™.Borosilicate Dr. Schilling Burette.

Specifications

Color Brown
Material Borosilicate Glass
Height (Metric) 900 mm
Includes PE-LD Reservoir Bottle
Stopcock Material PMP, PTFE
View More Specs

Description

Description

  • Patented VITLAB™ symbiotic (DE 10 2005 034 963) impact protection
  • Thermally stable plastic coating of the glass burette tube
  • Provides additional breaking resistance and acts as a splinter protection
Specifications

Specifications

Brown
900 mm
PMP, PTFE
Class B
Borosilicate Glass
PE-LD Reservoir Bottle
Schilling Burette
DIN ISO 384 Class B
Videos
SDS
Documents

Documents

Product Certifications