Learn More
Emma Safety Footwear Boston Safety Shoes
The TPU GripForce™ Easy Twist outsole offers maximum grip, is durable and highly recyclable. The integrated lacing prevents pressure points from hooks and eyes and the Hydro-Tec™ lining provides optimal breathability.
Brand:  Emma Safety Footwear MM40064747
 View more versions of this product
Product Code. 15958391
  • 116.00 € / 1 pair
Estimated Shipment: 24-02-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • S1P-Safety shoe with sole plate for dry work
  • Knitted Textile construction
  • Polyurethane coated
  • Hydro-Tec™ Antibacterial
  • Metal-free
  • Chrome-free lining
  • ESD Certified
Specifications
Specifications
EN ISO 20345:2011
Black, Gray
Hydro-Tec™ Antibacterial
47
Laces
Boston
Textile
Composite