missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Saint-Gobain Chemware™ PFA, Carboy
Closure Material: PTFE
Brand: Saint-Gobain D1069731
This item is not returnable.
View return policy
Description
- Chemically inert and ultra-pure; meets USP Class VI criteria
- Lowest metal ion content of all fluoropolymers
- Ideal for storage during intermediate phases of pharmaceutical and semiconductor processing

Specifications
5.3 gal. | |
PTFE | |
Closure | |
Carboy |
20 L | |
40.5 mm | |
PFA |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction