Learn More
Corning™ Bottle Top Dispenser (BTD) PTFE Plunger
PTFE plunger for BTD and dispensing reagents
Brand: Corning™ 6866
This item is not returnable.
View return policy
Description
- Specialized airpurging and closed circuit system ensures no loss of reagent during dispensing, as well as when the device is not in use
- Comes with four adapters with different diameters for universal use with multiple bottle sizes
- Telescopic suction tube can be adjusted smoothly to different bottle heights
- Calibration tool allows easy in-lab calibration with built-in recalibration mechanism to ensure the highest reproducibility during the dispenser’s long life cycle
- Every dispenser is individually tested and delivered with a certificate of performance
- For dispensing organic and inorganic liquids
Specifications
100 mL | |
Dispensing Organic and Inorganic Liquids | |
Four adapters |
Product Content Correction
Product Title
Corning™ Bottle Top Dispenser (BTD) PTFE Plunger > 100 mL
Spot an opportunity for improvement?Share a Content Correction