missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
- Supplied with tamper evident polypropylene cap
- Color: Clear
- Winchester pattern
- Unapproved

Specifications
Specifications
Capacity (Metric) | 1 L |
Color | Clear |
Diameter (Metric) Outer Neck | 45 mm |
Material | HDPE |
Quantity | 4 |
Width (Metric) | 100 |
Certifications/Compliance | Tested and certified to meet UN standards for the safe transportation of hazardous materials |
Mouth | Wide |
Height (Metric) | 210 mm |
Closure Material | Polypropylene |
Show More |
Documents
Scientific Resources
Brochures
Fisherbrand Focus on glassware and plasticware
File size: 66.4 MB
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Fisherbrand™ HDPE Wide Neck 1L Bottle >
Spot an opportunity for improvement?Share a Content Correction