missing translation for 'onlineSavingsMsg'
Learn More

Memmert™ Bottom Drip Tray

Material: Stainless Steel; For Use With: UF450

Brand:  Memmert™ U450B04362

Product Code. 15826311

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »

Explore more special offers
This item is not returnable. View return policy

Description

Description

  • May affect temperature distribution
  • Constructed of stainless steel
  • Rim height: 15 mm
  • Max. loading per tray: 8 kg
Specifications

Specifications

Up to 8 kg
Stainless Steel
Drip Tray
15 mm rim
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers