Learn More
Invitrogen™ Bovine anti-Mouse IgG (H+L) Secondary Antibody
Bovine Polyclonal Secondary Antibody
Brand: Invitrogen™ SA19502
100.67 EUR valid until 2025-03-29
Use promo code "24111" to get your promotional price.
Alert:
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”24111” to get your promotional price
Description
This antibody reacts with the heavy and light chains of the immunoglobulin molecule. SA1-9502 immunogen is purified mouse IgG.
Anti-Mouse secondary antibodies are affinity-purified antibodies with well-characterized specificity for mouse immunoglobulins and are useful in the detection, sorting or purification of its specified target. Secondary antibodies offer increased versatility enabling users to use many detection systems (e.g. HRP, AP, fluorescence). They can also provide greater sensitivity through signal amplification as multiple secondary antibodies can bind to a single primary antibody. Most commonly, secondary antibodies are generated by immunizing the host animal with a pooled population of immunoglobulins from the target species and can be further purified and modified (i.e. immunoaffinity chromatography, antibody fragmentation, label conjugation, etc.) to generate highly specific reagents.
Specifications
Mouse IgG (H+L) | |
Polyclonal | |
Unconjugated | |
Bovine | |
Affinity chromatography | |
RUO | |
Mouse | |
Secondary Antibody | |
IgG |
ELISA, Western Blot | |
1 mg/mL | |
PBS with 0.02% sodium azide | |
Purified mouse IgG. | |
500 μg | |
Secondary | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.