missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ Bovine Serum Albumin

Blocking buffer for IHC, ICC, ELISA and Western blotting; used to block non-specific antibody binding

Brand:  Tocris Bioscience™ 5217/100G

215.11 EUR valid until 2024-12-16
Use promo code "21630" to get your promotional price.


Product Code. 18776539

  • 281.00 € / 100g

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

Bovine Serum Albumin is a BSA (bovine serum albumin) is a widely used blocking reagent for use in immunohistochemistry (IHC), Immunocytochemistry (ICC), ELISAs and Western blotting. Used to block non-specific binding of antibodies. Can also be used as a protein concentration standard in Bradford assay for protein quantification. BSA is also used in cell culture.
Specifications

Specifications

100 g
Bovine Serum Albumin
0.98
Store at +4°C
Bovine
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

For Research Use Only