missing translation for 'onlineSavingsMsg'
Learn More

Scotch™ Box Sealing Tape 311+

This high-performance industrial packaging tape is for box sealing, splicing, recouperage and other demanding packaging applications. It offers good performance in damp and cold shipping and storage environments, even in temperatures below freezing.

Brand:  Scotch™ 7100220358

269.16 EUR valid until 2025-02-13
Use promo code "24130" to get your promotional price.


 View more versions of this product

Product Code. 17721679

  • 298.00 € / 6 rolls
Estimated Shipment: 25-02-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • High-performance industrial packaging tape designed for cold and damp temperatures
  • Adheres to highly recycled, corrugated boxes
  • Good performance over broad temperature ranges
  • Quick and easy application by hand, using manual dispensers or using 3M-Matic™ Case Sealers
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Pressure-sensitive Acrylic
Tape
Box Sealing Tape
3248 ft.
1.88 in.
Transparent
Polypropylene
990 m x 48 mm
990 m
48 mm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.