Learn More
Trajan™ BP1 GC Capillary Columns
Suitable for analysis of hydrocarbons, aromatics, pesticides, phenol, herbicides, and amines. Trajan Scientific and Medical™ BP1 CG Capillary Columns are constructed of 100% Dimethyl Polysiloxane.
Brand:  Trajan™ 054051
 View more versions of this product
Product Code. 16672602
  • 1065.00 € / Each
Estimated Shipment: 18-02-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
Suitable for analysis of hydrocarbons, aromatics, pesticides, phenol, herbicides, and amines. Trajan Scientific and Medical™ BP1 CG Capillary Columns are constructed of 100% Dimethyl Polysiloxane.
  • Classic cross-linked dimethyl polysiloxane technology
  • Low bleed
  • Non-polar
  • Ideal for analyses such as hydrocarbons, aromatics, pesticides and amines
Specifications
Specifications
340/360°C
0.22 mm
G1, G2, G38
-60°C to +340/360°C
60 m
Non polar
Van der Waals interaction
100% Dimethyl Polysiloxane
Phenols Analysis, Aromatics Analysis, Pesticide Analysis, Phenols, Herbicides Analysis, Amines
0.25 μm
Forte™ BP1
Fused Silica