missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BQ 788 sodium salt, Tocris Bioscience™
Click to view available options
Quantity:
1 mg
Description
Potent, selective ETB receptor antagonist (IC50 = 1.2 nM for inhibition of ET-1 binding to human Girardi heart cells).

Specifications
Specifications
Molecular Formula | C34H50N5NaO7 |
Quantity | 1 mg |
Synonym | N-[(cis-2,6-Dimethyl-1-piperidinyl)carbonyl]-4-methyl-L-leucyl-1-(methoxycarbonyl)-D-tryptophyl-D-norleucine sodium salt |
Formula Weight | 663.79 |
Percent Purity | >98% |
Physical Form | Solid |
Chemical Name or Material | BQ 788 sodium salt |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction