Learn More
Brady™ BBP™ Blank Yellow Pre-Cut Blank Sign Headers
BBP™ Blank Yellow Pre-Cut Blank Sign Headers are made of durable, low-shrink vinyl with ultra-aggressive adhesive that is ideal for labeling pipes, equipment, shelves and nearly any other industrial surface.
Brand:  Brady™ 142199
 View more versions of this product
Product Code. 11787324
  • 295.00 € / Each
Estimated Shipment: 15-01-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Highly pliable material with high-tack permanent adhesive for rough, highly textured, dirty and harsh environment surfaces—including outdoors
  • Conforms and adheres to "difficult" surfaces like painted cinder blocks, painted wood, textured plastic, paper-jacketed pipes and blow-molded cases
  • Resistant to exposure to chemicals, greases and oils, salt spray, cleaners and detergents and general industrial grime
  • 8-10 year average outdoor durability in temperatures from -40°F to 180°F (-40°C to 82°C); can be applied as low as 0°F (-18°C)
Specifications
Specifications
ASME ANSI Z535, OSHA 1910.145
Outdoor Grade Vinyl Labels for BBP3x Printers
Yellow
Tape
Non-sterile
BBP30, BBP31, BBP33, BBP35, and BBP37 printers
1
300/Pk.
-40 to 179.6°F
0.004 in.
82.60 mm
76.2 mm
5.71 cm
142199
Acrylic, Permanent
Gloss
Vinyl
Pipe marking, arrow tape, warehouse marking, warning panels, safety signs, arc flash and visual workplace labels
50 Gardner Units
8 to 10 Years
Brady Workstation, LabelMark, MarkWare
-40°C to +82°C
0.102 mm
2.25 x 3 in.
2.25 in.