Learn More
Brady™ BBP™ NOTICE Pre-Printed Pre-Cut Blank Sign Headers
BBP™ NOTICE Pre-Printed Pre-Cut Blank Sign Headers are made of durable, low-shrink vinyl with ultra-aggressive adhesive that is ideal for labeling pipes, equipment, shelves and nearly any other industrial surface.
Brand:  Brady™ 142188
 View more versions of this product
Product Code. 11737254
  • 457.00 € / Each
Estimated Shipment: 20-02-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Pre-printed headers result in multicolor header signs using a single, black print pass
  • Highly pliable material with high-tack permanent adhesive for rough, highly textured, dirty and harsh environment surfaces—including outdoors
  • Conforms and adheres to "difficult" surfaces like painted cinder blocks, painted wood, textured plastic, paper-jacketed pipes and blow-molded cases
  • Resistant to exposure to chemicals, greases and oils, salt spray, cleaners and detergents and general industrial grime
Specifications
Specifications
ASME ANSI Z535, OSHA 1910.145
Gloss
Vinyl
Permanent Acrylic
Pipemarkers, arrow tape, warehouse marking, warning panels, safety signs, arc flash and visual workplace labels
152 mm
-40°F
6 in.
-40°C to 82°C
English
Notice
Blue on White
B30-25-595-ANSINO
BBP30, BBP31, BBP33, BBP35, and BBP37 printers
175/Pk.
Pre-Printed Pre-Cut Blank Sign headers for BBP3x Printers
4 in.
180°F
0.004 in.
152.4 mm
-40°F to 180 °F
101 x 152 mm
Indoor/Outdoor Tape