Learn More
Brady™ BBP™ Repositional Vinyl Film
BBP™ Repositional Vinyl Film (B-581) is a durable, flexible, high gloss material ideal for indoor marking where frequent repositioning is required.
128.00 € - 457.00 €
Specifications
Adhesive Type Acrylic, Repositionable, Pressure-sensitive
Finish Gloss
Format Film
Material Vinyl
Sterility Non-sterile
View More Specs
Products 8
Product Code Brand Color Width (English) Price Quantity & Availability  
Product Code Brand Color Width (English) Price Quantity & Availability  
11706465
View Documents
Brady™
118003
White 4 in.
457.00 €
Each
Estimated Shipment: 30-01-2025
to see stock.
Add to basket
11716465
View Documents
Brady™
118004
White 2.25 in.
305.00 €
Each
Estimated Shipment: 30-01-2025
to see stock.
Add to basket
11736465
View Documents
Brady™
118006
White 0.5 in.
128.00 €
Each
Estimated Shipment: 27-02-2025
to see stock.
Add to basket
11756465
View Documents
Brady™
118008
Yellow 2.25 in.
305.00 €
Each
Estimated Shipment: 30-01-2025
to see stock.
Add to basket
11726465
View Documents
Brady™
118005
White 1.12 in.
204.00 €
Each
Estimated Shipment: 30-01-2025
to see stock.
Add to basket
11766465
View Documents
Brady™
118009
Yellow 1.12 in.
204.00 €
Each
Estimated Shipment: 27-02-2025
to see stock.
Add to basket
11746465
View Documents
Brady™
118007
Yellow 4 in.
457.00 €
Each
Estimated Shipment: 30-01-2025
to see stock.
Add to basket
11776465
View Documents
Brady™
118010
Yellow 0.5 in.
128.00 €
Each
Estimated Shipment: 27-02-2025
to see stock.
Add to basket
Description
Description
  • Special aggressive adhesive stays in place but removes cleanly without leaving adhesive residue making it ideal for applications like 5S and inventory
  • Vinyl film can be thermal transfer printed
  • Average outdoor durability is 5 years
  • Service temperature of -40°F to 180°F (-40°C to 82°C)
Specifications
Specifications
Acrylic, Repositionable, Pressure-sensitive
Film
Non-sterile
BBP30, BBP31, BBP33, BBP35, and BBP37 printers
1 Roll
-40°C to +82°C
100 ft.
Gloss
Vinyl
Ideal for temporary, Removable and repositionable labels, like 5S and lean inventory label
5 Years
-40 to 180°F
0.005 in.
30.48 m