Learn More
Brady™ BMP™ 21 ID Pal™ Label Cartridges: Self Laminating Vinyl
Self-laminating vinyl thermal transfer labels.
57.70 € - 72.00 €
Specifications
Autoclavable Not Autoclavable
Certifications/Compliance UL
Description Self-laminating Vinyl tape for M210, M210-LAB, M211, BMP21-PLUS, BMP21-LAB
Adhesive Type Permanent Acrylic, Permanent
Color White, Translucent, Black
View More Specs
Products 3
Product Code Brand Width (English) Price Quantity & Availability  
Product Code Brand Width (English) Price Quantity & Availability  
11543894
View Documents
Brady™
110928
1 in.
69.80 €
Each
In Stock
to see stock.
Add to basket
11563894
View Documents
Brady™
110930
1.5 in.
72.00 €
Each
In Stock
to see stock.
Add to basket
11533894
View Documents
Brady™
110927
0.75 in.
57.70 €
Each
In Stock
to see stock.
Add to basket
Description
Description
  • Sticks well to both smooth and textured surfaces.
  • Allows you to label cables and wires with a simple wrapping function.
  • Provides excellent smudge resistant text.
Specifications
Specifications
Not Autoclavable
Self-laminating Vinyl tape for M210, M210-LAB, M211, BMP21-PLUS, BMP21-LAB
White, Translucent, Black
Cartridge (Label)
Label Cartridge
M210, M210-LAB, M211, BMP21-PLUS, BMP21-LAB, IDPAL, LABPAL
-70°C
Rectangle
-70°C to +70°C
14 ft.
14 ft.
UL
Permanent Acrylic, Permanent
Matte
Vinyl
Non-sterile
M210 Label Printer, M210-LAB Label Printer, M211 Label Printer, BMP™21 LAB Label Printer, BMP™21-PLUS Label Printer
1 Roll
-94°F to +158°F
0.102 mm
4.30 m
4.2 m