missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Brady™ BMP51/BMP53/BMP41 Label Maker Cartridge: B-428 Metalized Polyester
For marking relays, solenoids, switches, drive motors and other components
Brand: Brady™ 711206
This item is not returnable.
View return policy
Description
- Available in various sizes
- For use with BMP41, BMP51, BMP53 printers
Specifications
Pass 2002/95/EC | |
Metallized Polyester (B-428) | |
239/Pk. | |
28.7 x 12.7 mm | |
28.7 mm | |
12.7 mm |
Black, Light Gray | |
Label Maker Cartridge | |
1.13 x 0.5 in. | |
1.13 in. | |
0.5 in. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction