Learn More
Brady™ FreezerBondz™ Ultra Thin Cryogenic Polyester Labels for B33 Printers
Durable Freezerbondz™ Polyester (B-492) markers are designed for laboratory identification applications such as vials, centrifuge, tubes, test tubes, straws and slides.
Brand:  Brady™ 143067
228.95 EUR valid until 2024-12-20
Use promo code "21731" to get your promotional price.
 View more versions of this product
Product Code. 13593310
  • 248.00 € / Each
In Stock
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Specialized — Aggressive adhesive B-492 FreezerBondz™ labels are ideal for curved, frozen surfaces like vials, centrifuge samples, tubes, and slides.
  • Durable — Labels withstand chemicals, abrasions, storage in liquid nitrogen, and a high/low temperature range of -320°F to +212°F (-196°C to +100°C).
  • Adhesive — Permanent acrylic adhesive for tight bonding onto smooth, curved surfaces, or wrapping around tubes and vials with a 1/8 in. overlap.
  • Certifications — RoHS compliant to 2011/65/EU (Pass 2015/863).
Specifications
Specifications
Not Autoclavable
FreezerBondz Labels for the BBP33 Printer
White
Polyester
Non-sterile
BBP33, i3300, i5300
1
LabelMark, MarkWare
-195 to 130°C
0.081 mm
1 x 1 in.
25.40 mm
2.5 cm
RoHS
Permanent Acrylic, Permanent
Matte
Laboratory Label
Use in laboratory identification such as vials, centrifuge tubes, test tubes, straws and slides
Cartridge of 1900 Labels
1900/Roll
-320°F to +230°F
0.0032 in.
27.94 mm
1 in.
1 in.