missing translation for 'onlineSavingsMsg'
Learn More

Brady™ IDXPERT™ and LABXPERT™ Labels

Tested to stand up to toughest, most extreme conditions

63.20 € - 130.00 €

Specifications

Product Type Label
For Use With (Equipment) iDXPERT™, LabXpert™ Printer
View More Specs

Products 3
Product Code Brand Color Quantity Height (Metric) Material Price Quantity & Availability  
Product Code Brand Color Quantity Height (Metric) Material Price Quantity & Availability  
11841633
View Documents
Brady™
142325
White, Blue 1 Roll 1.2 cm Vinyl N/A N/A N/A
11831643
View Documents
Brady™
142314
White, Red 1 Roll 3.8 cm Vinyl N/A N/A N/A
10317752
View Documents
Brady™
060951
Black, White 350/Pk. 2.2 cm Polyester N/A N/A N/A
Description

Description

  • Won't fall off, crack or peel
  • Preserve the integrity of your most important specimens indefinitely
  • Designed to withstand extreme temperatures and hold up in harsh chemicals such as, Xylene, Ethanol, DMSO and IPA
  • Can be used with frozen surfaces including glass and polypropylene
  • Suitable in autoclave, freezer, hot water baths (100°C), liquid nitrogen environments
  • Compatible with LABXPERT/IDXPERT printer
  • Black on white/translucent
  • Matte finish with smooth surface
  • Low-profile materials will not get stuck in centrifuge or other equipment

Order Info

For use with Brady LABXPERT and IDXPERT printers

Specifications

Specifications

Label
iDXPERT™, LabXpert™ Printer
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Brady™ IDXPERT™ and LABXPERT™ Labels

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.